Anti-PHYH Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: PHYH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77794-075
- 77794-076
- Antigen Symbol:
- PHYH
- PHYH
- Antigen Name:
- Phytanoyl-CoA dioxygenase, peroxisomal protein (1-338AA)
- Phytanoyl-CoA dioxygenase, peroxisomal protein (1-338AA)
- Antigen Synonyms:
- LN1|LNAP1|LNAP1, mouse, homolog of|OTTHUMP00000019131|OTTHUMP00000019132|OTTHUMP00000179083|OTTHUMP00000216226|PAHX|PAHX_HUMAN|peroxisomal|PhyH|PHYH1|Phytanic acid oxidase|phytanoil-CoA alpha hydroxylase|phytanoyl CoA 2 hydroxylase|Phytanoyl CoA 2 oxoglutarate dioxygenase|Phytanoyl CoA alpha hydroxylase|Phytanoyl CoA dioxygenase|Phytanoyl CoA dioxygenase peroxisomal|Phytanoyl-CoA alpha-hydroxylase|Phytanoyl-CoA dioxygenase|RD
- LN1|LNAP1|LNAP1, mouse, homolog of|OTTHUMP00000019131|OTTHUMP00000019132|OTTHUMP00000179083|OTTHUMP00000216226|PAHX|PAHX_HUMAN|peroxisomal|PhyH|PHYH1|Phytanic acid oxidase|phytanoil-CoA alpha hydroxylase|phytanoyl CoA 2 hydroxylase|Phytanoyl CoA 2 oxoglutarate dioxygenase|Phytanoyl CoA alpha hydroxylase|Phytanoyl CoA dioxygenase|Phytanoyl CoA dioxygenase peroxisomal|Phytanoyl-CoA alpha-hydroxylase|Phytanoyl-CoA dioxygenase|RD
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14832
- O14832
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice