Anti-USP4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77822-367
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 4 protein (632-810AA)
- Antigen Symbol:USP4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13107
- Antigen Synonyms:Deubiquitinating enzyme 4|MGC149848|MGC149849|Proto oncogene|Protooncogene|Ubiquitin carboxyl terminal hydrolase 4|Ubiquitin carboxyl-terminal hydrolase 4|Ubiquitin specific peptidase 4 (proto oncogene)|Ubiquitin specific peptidase 4 (protooncogene)|Ubiquitin specific peptidase 4|Ubiquitin specific processing protease 4|Ubiquitin specific protease 4 (proto oncogene)|Ubiquitin specific protease 4 (protooncogene)|Ubiquitin specific protease 4|Ubiquitin specific protease proto oncogene|Ubiquitin specific protease protooncogene|Ubiquitin thioesterase 4|Ubiquitin thiolesterase 4|Ubiquitin-specific-processing protease 4|Ubiquitous nuclear protein homolog|UBP4_HUMAN|UNP|UNPH|USP 4|USP4
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:200
Type:
Antigen: USP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human