Anti-TSKU Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: TSKU
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77811-612
- 77803-612
- Antigen Symbol:
- TSKU
- TSKU
- Antigen Name:
- Tsukushin protein (77-353AA)
- Tsukushin protein (77-353AA)
- Antigen Synonyms:
- TSKU|E2IG4|LRRC54|TSK|UNQ850/PRO1788Tsukushin|Tsukushi|E2-induced gene 4 protein|Leucine-rich repeat-containing protein 54
- TSKU|E2IG4|LRRC54|TSK|UNQ850/PRO1788Tsukushin|Tsukushi|E2-induced gene 4 protein|Leucine-rich repeat-containing protein 54
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WUA8
- Q8WUA8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice