Anti-STEAP3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STEAP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-095
- 77828-096
- Antigen Symbol:
- STEAP3
- STEAP3
- Antigen Name:
- Metalloreductase STEAP3 protein (1-207AA)
- Metalloreductase STEAP3 protein (1-207AA)
- Antigen Synonyms:
- 1010001D01Rik|Dudlin 2|Dudlin2|Dudulin 2|Dudulin-2|Dudulin2|hpHyde|hTSAP6|Metalloreductase STEAP3|MGC93147|pHyde|Six transmembrane epithelial antigen of prostate 3|Six transmembrane prostate protein 3|Six-transmembrane epithelial antigen of prostate 3|STEA 3|STEA3_HUMAN|STEAP 3|STEAP family member 3|STEAP family member 3 metalloreductase|STEAP3|STMP 3|STMP3|TSAP 6|TSAP6|Tumor suppressor activated pathway protein 6|Tumor suppressor pHyde|Tumor suppressor-activated pathway protein 6
- 1010001D01Rik|Dudlin 2|Dudlin2|Dudulin 2|Dudulin-2|Dudulin2|hpHyde|hTSAP6|Metalloreductase STEAP3|MGC93147|pHyde|Six transmembrane epithelial antigen of prostate 3|Six transmembrane prostate protein 3|Six-transmembrane epithelial antigen of prostate 3|STEA 3|STEA3_HUMAN|STEAP 3|STEAP family member 3|STEAP family member 3 metalloreductase|STEAP3|STMP 3|STMP3|TSAP 6|TSAP6|Tumor suppressor activated pathway protein 6|Tumor suppressor pHyde|Tumor suppressor-activated pathway protein 6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q658P3
- Q658P3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice