Anti-IGSF11 Rabbit Recombinant Polyclonal Antibody
77827-100
New Product
- Antigen Name:Immunoglobulin superfamily member 11 protein (23-241AA)
- Antigen Symbol:IGSF11
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q5DX21
- Antigen Synonyms:IGSF11|BTIGSF|CXADRL1|VSIG3|Immunoglobulin superfamily member 11|Brain and testis-specific immunoglobulin superfamily protein|Bt-IGSF|V-set and immunoglobulin domain-containing protein 3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: IGSF11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse