Anti-MTAP Rabbit Recombinant Polyclonal Antibody
Catalog # 77824-717
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:S-methyl-5\\\'-thioadenosine phosphorylase protein (1-283AA)
- Antigen Symbol:MTAP
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13126
- Antigen Synonyms:5' methylthioadenosine phosphorylase|5''-methylthioadenosine phosphorylase|BDMF|c86fus|DMSFH|DMSMFH|Epididymis luminal protein 249|HEL 249|LGMBF|MeSAdo phosphorylase|Methylthioadenosine phosphorylase|MSAP|MTA phosphorylase|MTAP|MTAP_HUMAN|MTAPase|S methyl 5 thioadenosine phosphorylase|S methyl 5' thioadenosine phosphorylase|S-methyl-5''-thioadenosine phosphorylase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:2000 to 1:10000, IF: 1:50 to 1:200
Type:
Antigen: MTAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse