To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Synthesized peptide derived from MEK-3 around the non-phosphorylation site of S189.
- Antigen Symbol:MAP2K3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P46734
- Antigen Synonyms:AW212142|dual specificity mitogen activated protein kinase kinase 3|Dual specificity mitogen-activated protein kinase kinase 3|MAP kinase kinase 3|map2k3|MAPK ERK kinase 3|MAPK/ERK kinase 3|MAPKK 3|MAPKK3|MEK 3|MEK3|Mitogen activated protein kinase kinase 3|MKK 3|MKK3|mMKK3b|MP2K3_HUMAN|PRKMK 3|PRKMK3|protein kinase, mitogen-activated, kinase 3|SAPK kinase 2|SAPKK 2|SAPKK2|Stress activated protein kinase kinase 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: MAP2K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat