Anti-RGS2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RGS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-210
- 77797-209
- Antigen Symbol:
- RGS2
- RGS2
- Antigen Name:
- Regulator of G-protein signaling 2 protein (1-211AA)
- Regulator of G-protein signaling 2 protein (1-211AA)
- Antigen Synonyms:
- Basic helix-loop-helix phosphoprotein G0S8|Cell growth inhibiting protein 31|Cell growth-inhibiting gene 31 protein|G0 to G1 switch regulatory 8 24kD|G0/G1 switch regulatory protein 8|G0S8|GOS8|OTTHUMP00000060765|Regulator of G protein signaling 2|Regulator of G protein signalling 2 24kD|Regulator of G-protein signaling 2|Regulators of G protein signaling XRGSVI|RGS 2|RGS2|RGS2_HUMAN
- Basic helix-loop-helix phosphoprotein G0S8|Cell growth inhibiting protein 31|Cell growth-inhibiting gene 31 protein|G0 to G1 switch regulatory 8 24kD|G0/G1 switch regulatory protein 8|G0S8|GOS8|OTTHUMP00000060765|Regulator of G protein signaling 2|Regulator of G protein signalling 2 24kD|Regulator of G-protein signaling 2|Regulators of G protein signaling XRGSVI|RGS 2|RGS2|RGS2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41220
- P41220
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice