Anti-DCAF1 Rabbit Polyclonal Antibody
77804-400
New Product
- Antigen Name:VPRBP
- Antigen Symbol:DCAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y4B6
- Antigen Synonyms:DCAF1|KIAA0800|RIP|VPRBPDDB1- and CUL4-associated factor 1|HIV-1 Vpr-binding protein|VprBP|Serine/threonine-protein kinase VPRBP|EC 2.7.11.1|Vpr-interacting protein
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: DCAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat