Anti-UBE3A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: UBE3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77811-899
- 77803-899
- Antigen Symbol:
- UBE3A
- UBE3A
- Antigen Name:
- Ubiquitin-protein ligase E3A protein (83-234AA)
- Ubiquitin-protein ligase E3A protein (83-234AA)
- Antigen Synonyms:
- ANCR|Angelman syndrome|AS|CTCL tumor antigen se37 2|CTCL tumor antigen se37-2|E6 AP|E6AP|E6AP ubiquitin protein ligase|E6AP ubiquitin-protein ligase|EPVE6AP|FLJ26981|HECT-type ubiquitin transferase E3A|HPVE6A|Human papilloma virus E6 associated protein Angelman syndrome|Human papilloma virus E6 associated protein|Human papillomavirus E6-associated protein|NY REN 54 antigen|Oncogenic protein associated protein E6 AP|Oncogenic protein associated protein E6AP|Oncogenic protein-associated protein E6-AP|Renal carcinoma antigen NY REN 54|Renal carcinoma antigen NY-REN-54|UBE 3A|Ube3a|UBE3A protein|UBE3A_HUMAN|Ubiquitin protein ligase E3A|Ubiquitin-protein ligase E3A
- ANCR|Angelman syndrome|AS|CTCL tumor antigen se37 2|CTCL tumor antigen se37-2|E6 AP|E6AP|E6AP ubiquitin protein ligase|E6AP ubiquitin-protein ligase|EPVE6AP|FLJ26981|HECT-type ubiquitin transferase E3A|HPVE6A|Human papilloma virus E6 associated protein Angelman syndrome|Human papilloma virus E6 associated protein|Human papillomavirus E6-associated protein|NY REN 54 antigen|Oncogenic protein associated protein E6 AP|Oncogenic protein associated protein E6AP|Oncogenic protein-associated protein E6-AP|Renal carcinoma antigen NY REN 54|Renal carcinoma antigen NY-REN-54|UBE 3A|Ube3a|UBE3A protein|UBE3A_HUMAN|Ubiquitin protein ligase E3A|Ubiquitin-protein ligase E3A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q05086
- Q05086
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice