Anti-EYA1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EYA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-440
- 77834-441
- Antigen Symbol:
- EYA1
- EYA1
- Antigen Name:
- Eyes absent homolog 1 protein (168-321AA)
- Eyes absent homolog 1 protein (168-321AA)
- Antigen Synonyms:
- BOP|BOR|BOS1|EYA transcriptional coactivator and phosphatase 1|Eya1|EYA1_HUMAN|Eyes absent 1|Eyes absent 1 homolog|Eyes absent homolog 1 (Drosophila)|Eyes absent homolog 1|Eyes absent homolog1|MGC141875|OFC1
- BOP|BOR|BOS1|EYA transcriptional coactivator and phosphatase 1|Eya1|EYA1_HUMAN|Eyes absent 1|Eyes absent 1 homolog|Eyes absent homolog 1 (Drosophila)|Eyes absent homolog 1|Eyes absent homolog1|MGC141875|OFC1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99502
- Q99502
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice