Anti-RMI1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: RMI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-518
- 77839-517
- Antigen Symbol:
- RMI1
- RMI1
- Antigen Name:
- RecQ-mediated genome instability protein 1 protein (181-443AA)
- RecQ-mediated genome instability protein 1 protein (181-443AA)
- Antigen Synonyms:
- BLAP75| BLM associated polypeptide 75 kDa| BLM associated protein 75 kDa| BLM-associated protein of 75 kDa| C9orf76| FAAP75| FLJ12888| Homolog of yeast RecQ mediated genome instability 1| RecQ mediated genome instability 1| RecQ-mediated genome instability protein 1| Rmi1| RMI1 homolog| RMI1 RecQ mediated genome instability 1 homolog| RMI1_HUMAN| RP11-346I8
- BLAP75| BLM associated polypeptide 75 kDa| BLM associated protein 75 kDa| BLM-associated protein of 75 kDa| C9orf76| FAAP75| FLJ12888| Homolog of yeast RecQ mediated genome instability 1| RecQ mediated genome instability 1| RecQ-mediated genome instability protein 1| Rmi1| RMI1 homolog| RMI1 RecQ mediated genome instability 1 homolog| RMI1_HUMAN| RP11-346I8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H9A7
- Q9H9A7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice