Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FAM96A Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-FAM96A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FAM96A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77838-400
  • 77838-399
  • Antigen Symbol:
  • FAM96A
  • FAM96A
  • Antigen Name:
  • Cytosolic iron-sulfur assembly component 2A protein (1-160AA)
  • Cytosolic iron-sulfur assembly component 2A protein (1-160AA)
  • Antigen Synonyms:
  • CIAO2A| CIA2A| FAM96A| Cytosolic iron-sulfur assembly component 2A| MIP18 family protein FAM96A
  • CIAO2A| CIA2A| FAM96A| Cytosolic iron-sulfur assembly component 2A| MIP18 family protein FAM96A
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H5X1
  • Q9H5X1
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications