Anti-R107.2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: R107.2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Caenorhabditis elegans
Specifications
- Catalog No:
- 77816-000
- 77816-001
- Antigen Symbol:
- R107.2
- R107.2
- Antigen Name:
- Recombinant Caenorhabditis elegans ATP-dependent (S)-NAD(P)H-hydrate dehydratase protein (1-307AA)
- Recombinant Caenorhabditis elegans ATP-dependent (S)-NAD(P)H-hydrate dehydratase protein (1-307AA)
- Antigen Synonyms:
- R107.2ATP-dependent|S)-NAD(P)H-hydrate dehydratase|EC 4.2.1.93|ATP-dependent NAD(P)HX dehydratase
- R107.2ATP-dependent|S)-NAD(P)H-hydrate dehydratase|EC 4.2.1.93|ATP-dependent NAD(P)HX dehydratase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- C. elegans
- C. elegans
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32740
- P32740
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together