Anti-SET Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: SET
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-285
- 77839-284
- Antigen Symbol:
- SET
- SET
- Antigen Name:
- Protein SET protein (147-235AA)
- Protein SET protein (147-235AA)
- Antigen Synonyms:
- 2PP2A|HLA DR associated protein II|HLA-DR-associated protein II|I 2PP2A|I-2PP2A|I2PP2A|IGAAD|Inhibitor of granzyme A activated DNase|Inhibitor of granzyme A-activated DNase|Inhibitor of GzmA-activated DNase|inhibitor-2 of protein phosphatase-2A|IPP2A2|PHAPII|Phosphatase 2A inhibitor I2PP2A|protein phosphatase type 2A inhibitor|Protein SET|Set|SET nuclear oncogene|SET translocation|SET translocation (myeloid leukemia-associated)|SET_HUMAN|TAF I|TAF IBETA|TAF-I|TAFI|Template activating factor I|Template-activating factor I|Template-Activating Factor-I, chromatin remodelling factor
- 2PP2A|HLA DR associated protein II|HLA-DR-associated protein II|I 2PP2A|I-2PP2A|I2PP2A|IGAAD|Inhibitor of granzyme A activated DNase|Inhibitor of granzyme A-activated DNase|Inhibitor of GzmA-activated DNase|inhibitor-2 of protein phosphatase-2A|IPP2A2|PHAPII|Phosphatase 2A inhibitor I2PP2A|protein phosphatase type 2A inhibitor|Protein SET|Set|SET nuclear oncogene|SET translocation|SET translocation (myeloid leukemia-associated)|SET_HUMAN|TAF I|TAF IBETA|TAF-I|TAFI|Template activating factor I|Template-activating factor I|Template-Activating Factor-I, chromatin remodelling factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q01105
- Q01105
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice