Anti-ACS8 Rabbit Recombinant Polyclonal Antibody
Catalog # 77837-834
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469AA)
- Antigen Symbol:ACS8
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:A. thaliana
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9T065
- Antigen Synonyms:ACS8|At4g37770|T28I19.501-aminocyclopropane-1-carboxylate synthase 8|ACC synthase 8|EC 4.4.1.14|S-adenosyl-L-methionine methylthioadenosine-lyase 8
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: ACS8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana