Anti-UGT2B10 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: UGT2B10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-974
- 77811-974
- Antigen Symbol:
- UGT2B10
- UGT2B10
- Antigen Name:
- UDP-glucuronosyltransferase 2B10 protein (20-280AA)
- UDP-glucuronosyltransferase 2B10 protein (20-280AA)
- Antigen Synonyms:
- MGC142209|UDB10_HUMAN|UDP glucuronosyltransferase 2 family polypeptide B10|UDP glucuronosyltransferase 2B10|UDP glycosyltransferase 2 family polypeptide B10|UDP-glucuronosyltransferase 2B10|UDPGT 2B10|UDPGT|UDPGT2B10|UGT2B10
- MGC142209|UDB10_HUMAN|UDP glucuronosyltransferase 2 family polypeptide B10|UDP glucuronosyltransferase 2B10|UDP glycosyltransferase 2 family polypeptide B10|UDP-glucuronosyltransferase 2B10|UDPGT 2B10|UDPGT|UDPGT2B10|UGT2B10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36537
- P36537
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice