Anti-ADAMTS15 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: ADAMTS15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77808-917
- 77808-918
- Antigen Symbol:
- ADAMTS15
- ADAMTS15
- Antigen Name:
- Fusion protein of ADAMTS15
- Fusion protein of ADAMTS15
- Antigen Synonyms:
- A disintegrin and metalloproteinase with thrombospondin motifs 15| A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15| A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15 preproprotein| A disintegrin like and metalloprotease with thrombospondin type 1 motif 15| A disintegrin like and metalloprotease with thrombospondin type 1 motif 15 preproprotein| ADAM metallopeptidase with thrombospondin type 1 motif 15| ADAM TS 15| ADAM TS15| ADAM-TS 15| ADAM-TS15| ADAMTS 15| ADAMTS-15| ADAMTS15| ATS15_HUMAN| MGC126403
- A disintegrin and metalloproteinase with thrombospondin motifs 15| A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15| A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15 preproprotein| A disintegrin like and metalloprotease with thrombospondin type 1 motif 15| A disintegrin like and metalloprotease with thrombospondin type 1 motif 15 preproprotein| ADAM metallopeptidase with thrombospondin type 1 motif 15| ADAM TS 15| ADAM TS15| ADAM-TS 15| ADAM-TS15| ADAMTS 15| ADAMTS-15| ADAMTS15| ATS15_HUMAN| MGC126403
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE58
- Q8TE58
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice