Anti-MSRB1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MSRB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-197
- 77841-198
- Antigen Symbol:
- MSRB1
- MSRB1
- Antigen Name:
- Methionine-R-sulfoxide reductase B1 protein (1-116AA)
- Methionine-R-sulfoxide reductase B1 protein (1-116AA)
- Antigen Synonyms:
- Annexin A2 like|EC 1 8 4|HSPC 270|HSPC270|Methionine R sulfoxide reductase|Methionine R sulfoxide reductase B1|Methionine sulfoxide reductase|Methionine-R-sulfoxide reductase B1|MGC3344|MSRB 1|MsrB1|MSRB1_HUMAN|OTTHUMP00000159418|Sel X|Selenoprotein R|Selenoprotein X 1|Selenoprotein X|Selenoprotein X1|SELR|SelX|SEPX 1|sepx1
- Annexin A2 like|EC 1 8 4|HSPC 270|HSPC270|Methionine R sulfoxide reductase|Methionine R sulfoxide reductase B1|Methionine sulfoxide reductase|Methionine-R-sulfoxide reductase B1|MGC3344|MSRB 1|MsrB1|MSRB1_HUMAN|OTTHUMP00000159418|Sel X|Selenoprotein R|Selenoprotein X 1|Selenoprotein X|Selenoprotein X1|SELR|SelX|SEPX 1|sepx1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZV6
- Q9NZV6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice