Anti-RNF31 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RNF31
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-060
- 77834-059
- Antigen Symbol:
- RNF31
- RNF31
- Antigen Name:
- E3 ubiquitin-protein ligase RNF31 protein (421-700AA)
- E3 ubiquitin-protein ligase RNF31 protein (421-700AA)
- Antigen Synonyms:
- E3 ubiquitin protein ligase RNF31|FLJ10111|FLJ23501|HOIL 1L interacting protein|HOIL-1-interacting protein|HOIL1L interacting protein|HOIP|RING finger protein 31|RNF31|RNF31_HUMAN|ZIBRA|Zinc in between ring finger ubiquitin associated domain|Zinc in-between-RING-finger ubiquitin-associated domain protein
- E3 ubiquitin protein ligase RNF31|FLJ10111|FLJ23501|HOIL 1L interacting protein|HOIL-1-interacting protein|HOIL1L interacting protein|HOIP|RING finger protein 31|RNF31|RNF31_HUMAN|ZIBRA|Zinc in between ring finger ubiquitin associated domain|Zinc in-between-RING-finger ubiquitin-associated domain protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96EP0
- Q96EP0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice