Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MBD5 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-MBD5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MBD5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77837-694
  • 77837-693
  • Antigen Symbol:
  • MBD5
  • MBD5
  • Antigen Name:
  • Methyl-CpG-binding domain protein 5 protein (623-840AA)
  • Methyl-CpG-binding domain protein 5 protein (623-840AA)
  • Antigen Synonyms:
  • 9430004D19Rik| AA536666| AI426407| C030040A15Rik| FLJ11113| FLJ30517| KIAA1461 | MBD5| MBD5_HUMAN| methyl CpG binding domain protein 5| Methyl CpG binding protein MBD5 | Methyl-CpG-binding domain protein 5| Methyl-CpG-binding protein MBD5| RP23 167N13.1
  • 9430004D19Rik| AA536666| AI426407| C030040A15Rik| FLJ11113| FLJ30517| KIAA1461 | MBD5| MBD5_HUMAN| methyl CpG binding domain protein 5| Methyl CpG binding protein MBD5 | Methyl-CpG-binding domain protein 5| Methyl-CpG-binding protein MBD5| RP23 167N13.1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9P267
  • Q9P267
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications