Anti-USP9X Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: USP9X
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-873
- 77833-872
- Antigen Symbol:
- USP9X
- USP9X
- Antigen Name:
- Probable ubiquitin carboxyl-terminal hydrolase FAF-X protein (2360-2484AA)
- Probable ubiquitin carboxyl-terminal hydrolase FAF-X protein (2360-2484AA)
- Antigen Synonyms:
- Deubiquitinating enzyme FAF X| Deubiquitinating enzyme FAF-X| DFFRX| Drosophila fat facets related X linked| FAF| Fafl| Fam| Fat facets homolog| Fat facets in mammals| Fat facets protein related X linked| Fat facets protein related, X-linked| Fat facets protein-related| hFAM| MRX99| Probable ubiquitin carboxyl terminal hydrolase FAF X| Probable ubiquitin carboxyl-terminal hydrolase FAF-X| Ubiquitin carboxyl-terminal hydrolase FAM| Ubiquitin specific peptidase 9 X linked| Ubiquitin specific peptidase 9, X-linked| Ubiquitin specific processing protease FAF X| Ubiquitin specific protease 9 X chromosome| Ubiquitin thioesterase FAF X| Ubiquitin thiolesterase FAF X| Ubiquitin thiolesterase FAF-X| Ubiquitin-specific protease 9| Ubiquitin-specific-processing protease FAF-X| USP9 (gene name)| Usp9x| USP9X_HUMAN| Uubiquitin specific protease 9, X chromosome (fat facets like Drosophila)| X chromosome| X-linked
- Deubiquitinating enzyme FAF X| Deubiquitinating enzyme FAF-X| DFFRX| Drosophila fat facets related X linked| FAF| Fafl| Fam| Fat facets homolog| Fat facets in mammals| Fat facets protein related X linked| Fat facets protein related, X-linked| Fat facets protein-related| hFAM| MRX99| Probable ubiquitin carboxyl terminal hydrolase FAF X| Probable ubiquitin carboxyl-terminal hydrolase FAF-X| Ubiquitin carboxyl-terminal hydrolase FAM| Ubiquitin specific peptidase 9 X linked| Ubiquitin specific peptidase 9, X-linked| Ubiquitin specific processing protease FAF X| Ubiquitin specific protease 9 X chromosome| Ubiquitin thioesterase FAF X| Ubiquitin thiolesterase FAF X| Ubiquitin thiolesterase FAF-X| Ubiquitin-specific protease 9| Ubiquitin-specific-processing protease FAF-X| USP9 (gene name)| Usp9x| USP9X_HUMAN| Uubiquitin specific protease 9, X chromosome (fat facets like Drosophila)| X chromosome| X-linked
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q93008
- Q93008
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice