Anti-BIK Rabbit Polyclonal Antibody
Catalog # 77807-464
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from NBK around the phosphorylation site of S35
- Antigen Symbol:BIK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q13323
- Antigen Synonyms:Apoptosis inducer NBK|BBC1|Bcl-2-interacting killer|BCL2 interacting killer|bhikhari|BIK|Bik-like killer protein|BIK_HUMAN|BIP 1|BiP1|BP 4|BP4|cb 60|NBK
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% Sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended Dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: BIK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human