Anti-LAIR2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: LAIR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77814-829
- 77814-828
- Antigen Symbol:
- LAIR2
- LAIR2
- Antigen Name:
- Synthetic peptide of LAIR2
- Synthetic peptide of LAIR2
- Antigen Synonyms:
- CD306|CD306 antigen|LAIR 2|LAIR-2|LAIR2_HUMAN|Leukocyte associated Ig like receptor 2|Leukocyte associated immunoglobulin like receptor 2|Leukocyte-associated immunoglobulin-like receptor 2|MGC71634|OTTHUMP00000068389|OTTHUMP00000068390
- CD306|CD306 antigen|LAIR 2|LAIR-2|LAIR2_HUMAN|Leukocyte associated Ig like receptor 2|Leukocyte associated immunoglobulin like receptor 2|Leukocyte-associated immunoglobulin-like receptor 2|MGC71634|OTTHUMP00000068389|OTTHUMP00000068390
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ISS4
- Q6ISS4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice