Anti-RHD Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: RHD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77797-271
- 77797-272
- Antigen Symbol:
- RHD
- RHD
- Antigen Name:
- Blood group Rh(D) polypeptide protein (32-76AA)
- Blood group Rh(D) polypeptide protein (32-76AA)
- Antigen Synonyms:
- Blood group Rh(D) polypeptide|Blood group--Rhesus system D polypeptide|CD240D|D antigen (DCS)|DIIIc|MGC165007|RH|Rh blood group antigen Evans|Rh blood group D antigen|Rh polypeptide 2|RH30|Rh4|RHCED|RHD_HUMAN|RhDCw|RHDel|RHDVA(TT)|Rhesus blood group D antigen allele DIII type 7|Rhesus D antigen|Rhesus system D polypeptide|RhII|RhK562-II|RhPI|RHPII|RHXIII
- Blood group Rh(D) polypeptide|Blood group--Rhesus system D polypeptide|CD240D|D antigen (DCS)|DIIIc|MGC165007|RH|Rh blood group antigen Evans|Rh blood group D antigen|Rh polypeptide 2|RH30|Rh4|RHCED|RHD_HUMAN|RhDCw|RHDel|RHDVA(TT)|Rhesus blood group D antigen allele DIII type 7|Rhesus D antigen|Rhesus system D polypeptide|RhII|RhK562-II|RhPI|RHPII|RHXIII
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02161
- Q02161
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice