Anti-AKR1C3 Rabbit Polyclonal Antibody
77810-362
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Synthesized peptide derived from the Internal region of DD3.
- Antigen Symbol:AKR1C3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P42330
- Antigen Synonyms:17 beta HSD 5|17 beta hydroxysteroid dehydrogenase type 5|17-beta-HSD 5|17-beta-hydroxysteroid dehydrogenase type 5|2-dihydrobenzene-1|2-diol dehydrogenase|20 alpha-hydroxysteroid dehydrogenase|20-alpha-hydroxysteroid dehydrogenase|3 alpha hydroxysteroid dehydrogenase type II|3-alpha-HSD type 2|3-alpha-HSD type II|brain|3-alpha-hydroxysteroid dehydrogenase type 2|AK1C3_HUMAN|AKR1 C3|Akr1c18|AKR1C3|Aldo keto reductase family 1 member C3|Aldo-keto reductase family 1 member C3|Chlordecone reductase|Chlordecone reductase homolog HAKRb|DD-3|DD3|DDH1|DDX|Dihydrodiol dehydrogenase 3|Dihydrodiol dehydrogenase type I|Dihydrodiol dehydrogenase X|HA1753|HAKRB|HAKRe|hluPGFS|HSD17B5|Indanol dehydrogenase|KIAA0119|PGFS|Prostaglandin F synthase|Testosterone 17-beta-dehydrogenase 5|Trans-1|Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase|Type IIb 3 alpha hydroxysteroid dehydrogenase
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:20000
Type:
Antigen: AKR1C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human