Anti-SUGCT Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: SUGCT
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77837-256
- 77837-255
- Antigen Symbol:
- SUGCT
- SUGCT
- Antigen Name:
- Succinate--hydroxymethylglutarate CoA-transferase protein (1-250AA)
- Succinate--hydroxymethylglutarate CoA-transferase protein (1-250AA)
- Antigen Synonyms:
- SUGCT| C7orf10| DERP13| Succinate--hydroxymethylglutarate CoA-transferase| EC 2.8.3.13| Dermal papilla-derived protein 13| SuccinylCoA:glutarate-CoA transferase
- SUGCT| C7orf10| DERP13| Succinate--hydroxymethylglutarate CoA-transferase| EC 2.8.3.13| Dermal papilla-derived protein 13| SuccinylCoA:glutarate-CoA transferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HAC7
- Q9HAC7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice