Anti-MKRN1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MKRN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-378
- 77841-379
- Antigen Symbol:
- MKRN1
- MKRN1
- Antigen Name:
- E3 ubiquitin-protein ligase makorin-1 protein (109-209AA)
- E3 ubiquitin-protein ligase makorin-1 protein (109-209AA)
- Antigen Synonyms:
- E3 ubiquitin-protein ligase makorin-1|FLJ21334|HGNC:7112|Makorin|Makorin ring finger protein 1|MKRN 1|MKRN1_HUMAN|OTTHUMP00000212117|OTTHUMP00000212118|OTTHUMP00000212121|OTTHUMP00000212124|Ring finger protein 1|RING finger protein 61|RNF61
- E3 ubiquitin-protein ligase makorin-1|FLJ21334|HGNC:7112|Makorin|Makorin ring finger protein 1|MKRN 1|MKRN1_HUMAN|OTTHUMP00000212117|OTTHUMP00000212118|OTTHUMP00000212121|OTTHUMP00000212124|Ring finger protein 1|RING finger protein 61|RNF61
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHC7
- Q9UHC7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice