Anti-ARAP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ARAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77834-260
- 77834-259
- Antigen Symbol:
- ARAP1
- ARAP1
- Antigen Name:
- Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1 protein (69-344AA)
- Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1 protein (69-344AA)
- Antigen Synonyms:
- 2410002L19Rik|ANK repeat and PH domain-containing protein 1|ARAP 1|Arap1|ARAP1_HUMAN|Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domains containing protein 1|Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domain containing protein 1|Arf-GAP with Rho-GAP domain|CENT D2|Centaurin delta 2|Centaurin delta2|Centaurin-delta-2|CENTD 2|CENTD2|Cnt d2|Cnt-d2|Cntd2|KIAA0782|MGC176140|mKIAA0782
- 2410002L19Rik|ANK repeat and PH domain-containing protein 1|ARAP 1|Arap1|ARAP1_HUMAN|Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domains containing protein 1|Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domain containing protein 1|Arf-GAP with Rho-GAP domain|CENT D2|Centaurin delta 2|Centaurin delta2|Centaurin-delta-2|CENTD 2|CENTD2|Cnt d2|Cnt-d2|Cntd2|KIAA0782|MGC176140|mKIAA0782
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96P48
- Q96P48
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice