Anti-NSDHL Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NSDHL
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-698
- 77823-697
- Antigen Symbol:
- NSDHL
- NSDHL
- Antigen Name:
- Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating protein (154-211AA)
- Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating protein (154-211AA)
- Antigen Synonyms:
- decarboxylating|H105E3|H105e3 protein|NAD(P) dependent steroid dehydrogenase like|NSDHL|NSDHL_HUMAN|Protein H105e3|SDR31E1|Short chain dehydrogenase/reductase family 31E member 1|Sterol 4 alpha carboxylate 3 dehydrogenase decarboxylating|Sterol-4-alpha-carboxylate 3-dehydrogenase|XAP104
- decarboxylating|H105E3|H105e3 protein|NAD(P) dependent steroid dehydrogenase like|NSDHL|NSDHL_HUMAN|Protein H105e3|SDR31E1|Short chain dehydrogenase/reductase family 31E member 1|Sterol 4 alpha carboxylate 3 dehydrogenase decarboxylating|Sterol-4-alpha-carboxylate 3-dehydrogenase|XAP104
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15738
- Q15738
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together