Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AK3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-AK3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-AK3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77841-567
  • 77841-568
  • Antigen Symbol:
  • AK3
  • AK3
  • Antigen Name:
  • GTP:AMP phosphotransferase AK3, mitochondrial protein (142-227AA)
  • GTP:AMP phosphotransferase AK3, mitochondrial protein (142-227AA)
  • Antigen Synonyms:
  • Adenylate kinase 3 alpha-like 1| Adenylate kinase 3| Adenylate kinase 3, formerly| adenylate kinase 6, adenylate kinase 3 like 1| AK 3| AK3| AK3, formerly| AK3L1| AK4| AK4, mouse, homolog of| AK6| AKL3L| AKL3L1| FIX| GTP:AMP phosphotransferase| GTP:AMP phosphotransferase, mitochondrial| KAD3_HUMAN| mitochondrial| OTTHUMP00000021015| OTTHUMP00000021016| RP11 6J24.4
  • Adenylate kinase 3 alpha-like 1| Adenylate kinase 3| Adenylate kinase 3, formerly| adenylate kinase 6, adenylate kinase 3 like 1| AK 3| AK3| AK3, formerly| AK3L1| AK4| AK4, mouse, homolog of| AK6| AKL3L| AKL3L1| FIX| GTP:AMP phosphotransferase| GTP:AMP phosphotransferase, mitochondrial| KAD3_HUMAN| mitochondrial| OTTHUMP00000021015| OTTHUMP00000021016| RP11 6J24.4
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UIJ7
  • Q9UIJ7
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications