Anti-TBX4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77802-032
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:T-box transcription factor TBX4 protein (355-511AA)
- Antigen Symbol:TBX4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P57082
- Antigen Synonyms:SPS|T box 4|T box protein 4|T box transcription factor TBX 4|T box transcription factor TBX4|T-box protein 4|T-box transcription factor TBX4|TBX 4|TBX4|TBX4_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: TBX4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human