Anti-TINAGL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TINAGL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-307
- 77839-306
- Antigen Symbol:
- TINAGL1
- TINAGL1
- Antigen Name:
- Tubulointerstitial nephritis antigen-like protein (250-467AA)
- Tubulointerstitial nephritis antigen-like protein (250-467AA)
- Antigen Synonyms:
- TINAGL1|GIS5|LCN7|OLRG2|TINAGL|PP6614|PSEC0088|UNQ204/PRO230|Tubulointerstitial nephritis antigen-like|Glucocorticoid-inducible protein 5|Oxidized LDL-responsive gene 2 protein|OLRG-2|Tubulointerstitial nephritis antigen-related protein|TIN Ag-related protein|TIN-Ag-RP
- TINAGL1|GIS5|LCN7|OLRG2|TINAGL|PP6614|PSEC0088|UNQ204/PRO230|Tubulointerstitial nephritis antigen-like|Glucocorticoid-inducible protein 5|Oxidized LDL-responsive gene 2 protein|OLRG-2|Tubulointerstitial nephritis antigen-related protein|TIN Ag-related protein|TIN-Ag-RP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZM7
- Q9GZM7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice