Anti-USP12 Rabbit Recombinant Polyclonal Antibody
77804-104
New Product
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 12 protein (1-370AA)
- Antigen Symbol:USP12
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O75317
- Antigen Synonyms:Deubiquitinating enzyme 12|OTTHUMP00000042337|UBH 1|UBH1|Ubiquitin carboxyl terminal hydrolase 12|Ubiquitin carboxyl-terminal hydrolase 12|Ubiquitin hydrolyzing enzyme 1|Ubiquitin specific peptidase 12|Ubiquitin specific processing protease 12|Ubiquitin specific protease 12 like 1|Ubiquitin thioesterase 12|Ubiquitin thiolesterase 12|Ubiquitin-hydrolyzing enzyme 1|Ubiquitin-specific-processing protease 12|UBP12_HUMAN|USP 12|Usp12|USP12L1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:200 to 1:1000
Type:
Antigen: USP12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse