Anti-MAP3K19 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: MAP3K19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77827-673
- 77827-674
- Antigen Symbol:
- MAP3K19
- MAP3K19
- Antigen Name:
- Mitogen-activated protein kinase kinase kinase 19 protein (45-168AA)
- Mitogen-activated protein kinase kinase kinase 19 protein (45-168AA)
- Antigen Synonyms:
- MAP3K19|RCK|YSK4Mitogen-activated protein kinase kinase kinase 19|EC 2.7.11.1|Regulated in COPD|protein kinase|SPS1/STE20-related protein kinase YSK4
- MAP3K19|RCK|YSK4Mitogen-activated protein kinase kinase kinase 19|EC 2.7.11.1|Regulated in COPD|protein kinase|SPS1/STE20-related protein kinase YSK4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q56UN5
- Q56UN5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice