Anti-SARS Rabbit Polyclonal Antibody
77798-658
New Product
- Antigen Name:SYSC_HUMAN
- Antigen Symbol:SARS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P49591
- Antigen Synonyms:C78314|cytoplasmic|EC 6.1.1.11|FLJ36399|sarS|Sars1|Serine tRNA ligase 1, cytoplasmic|Serine tRNA ligase|Serine--tRNA ligase|serine--tRNA ligase, cytoplasmic|SerRS|SERS|Seryl tRNA Ser/Sec synthetase|Seryl tRNA synthetase|Seryl tRNA synthetase cytoplasmic|Seryl-tRNA Ser/Sec synthetase|Seryl-tRNA synthetase|seryl-tRNA synthetase, cytoplasmic|Seryl-tRNA(Ser/Sec) synthetase|Strs|SYSC_HUMAN
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: SARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat