Anti-PDE8B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDE8B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-557
- 77819-558
- Antigen Symbol:
- PDE8B
- PDE8B
- Antigen Name:
- High affinity cAMP-specific and IBMX-insensitive 3\',5\'-cyclic phosphodiesterase 8Bprotein (18-110AA)
- High affinity cAMP-specific and IBMX-insensitive 3\',5\'-cyclic phosphodiesterase 8Bprotein (18-110AA)
- Antigen Synonyms:
- 3' 5' cyclic nucleotide phosphodiesterase 8B|3'5' cyclic nucleotide phosphodiesterase 8B|Cell proliferation-inducing gene 22 protein|FLJ11212|High affinity cAMP specific and IBMX insensitive 3' 5' cyclic phosphodiesterase 8B|High affinity cAMP specific and IBMX insensitive 3'5' cyclic phosphodiesterase 8B|High affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8B|HSPDE 8B|HsPDE8B|PDE 8B|PDE8B|PDE8B_HUMAN|Phosphodiesterase 8B|Phosphodiesterase8B|PIG22
- 3' 5' cyclic nucleotide phosphodiesterase 8B|3'5' cyclic nucleotide phosphodiesterase 8B|Cell proliferation-inducing gene 22 protein|FLJ11212|High affinity cAMP specific and IBMX insensitive 3' 5' cyclic phosphodiesterase 8B|High affinity cAMP specific and IBMX insensitive 3'5' cyclic phosphodiesterase 8B|High affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8B|HSPDE 8B|HsPDE8B|PDE 8B|PDE8B|PDE8B_HUMAN|Phosphodiesterase 8B|Phosphodiesterase8B|PIG22
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95263
- O95263
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice