Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ZSCAN32 Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-ZSCAN32 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ZSCAN32 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77805-462
  • 77805-463
  • Antigen Symbol:
  • ZSCAN32
  • ZSCAN32
  • Antigen Name:
  • Zinc finger and SCAN domain-containing protein 32 protein (6-248AA)
  • Zinc finger and SCAN domain-containing protein 32 protein (6-248AA)
  • Antigen Synonyms:
  • ZSCAN32|ZNF434|HCCS5|Zinc finger and SCAN domain-containing protein 32|Human cervical cancer suppressor gene 5 protein|HCCS-5|Zinc finger protein 434
  • ZSCAN32|ZNF434|HCCS5|Zinc finger and SCAN domain-containing protein 32|Human cervical cancer suppressor gene 5 protein|HCCS-5|Zinc finger protein 434
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9NX65
  • Q9NX65
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications