Anti-POLR3B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: POLR3B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-463
- 77842-462
- Antigen Symbol:
- POLR3B
- POLR3B
- Antigen Name:
- DNA-directed RNA polymerase III subunit RPC2 protein (831-939AA)
- DNA-directed RNA polymerase III subunit RPC2 protein (831-939AA)
- Antigen Synonyms:
- C128|DNA directed RNA polymerase III 127.6 kDa polypeptide|DNA directed RNA polymerase III subunit B|DNA-directed RNA polymerase III 127.6 kDa polypeptide|DNA-directed RNA polymerase III subunit B|DNA-directed RNA polymerase III subunit RPC2|FLJ10388|polr3b|Polymerase (RNA) III (DNA directed) polypeptide B|RNA polymerase III subunit C2|RNA polymerase III subunit RPC2|RPC2|RPC2_HUMAN
- C128|DNA directed RNA polymerase III 127.6 kDa polypeptide|DNA directed RNA polymerase III subunit B|DNA-directed RNA polymerase III 127.6 kDa polypeptide|DNA-directed RNA polymerase III subunit B|DNA-directed RNA polymerase III subunit RPC2|FLJ10388|polr3b|Polymerase (RNA) III (DNA directed) polypeptide B|RNA polymerase III subunit C2|RNA polymerase III subunit RPC2|RPC2|RPC2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NW08
- Q9NW08
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice