Anti-ENKD1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ENKD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-077
- 77837-076
- Antigen Symbol:
- ENKD1
- ENKD1
- Antigen Name:
- Enkurin domain-containing protein 1 protein (1-346AA)
- Enkurin domain-containing protein 1 protein (1-346AA)
- Antigen Synonyms:
- Chromosome 16 open reading frame 48|DAKV6410|DKFZp434A1319|Enkd1|ENKD1_HUMAN|Enkurin domain containing 1|Enkurin domain-containing protein 1|Hypothetical protein LOC84080|UNQ6410
- Chromosome 16 open reading frame 48|DAKV6410|DKFZp434A1319|Enkd1|ENKD1_HUMAN|Enkurin domain containing 1|Enkurin domain-containing protein 1|Hypothetical protein LOC84080|UNQ6410
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H0I2
- Q9H0I2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice