Anti-USP10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-035
- 77822-036
- Antigen Symbol:
- USP10
- USP10
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 10 protein (453-592AA)
- Ubiquitin carboxyl-terminal hydrolase 10 protein (453-592AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 10|KIAA0190|MGC124997|MGC2621|Ubiquitin carboxyl terminal hydrolase 10|Ubiquitin carboxyl-terminal hydrolase 10|Ubiquitin specific peptidase 10|Ubiquitin specific processing protease 10|Ubiquitin specific protease 10|Ubiquitin thioesterase 10|Ubiquitin thiolesterase 10|Ubiquitin-specific-processing protease 10|UBP10_HUMAN|UBPO|USP 10|USP10
- Deubiquitinating enzyme 10|KIAA0190|MGC124997|MGC2621|Ubiquitin carboxyl terminal hydrolase 10|Ubiquitin carboxyl-terminal hydrolase 10|Ubiquitin specific peptidase 10|Ubiquitin specific processing protease 10|Ubiquitin specific protease 10|Ubiquitin thioesterase 10|Ubiquitin thiolesterase 10|Ubiquitin-specific-processing protease 10|UBP10_HUMAN|UBPO|USP 10|USP10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14694
- Q14694
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice