Anti-MRPL20 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPL20
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-953
- 77835-952
- Antigen Symbol:
- MRPL20
- MRPL20
- Antigen Name:
- 39S ribosomal protein L20, mitochondrial protein (46-149AA)
- 39S ribosomal protein L20, mitochondrial protein (46-149AA)
- Antigen Synonyms:
- MRPL20|39S ribosomal protein L20|mitochondrial|L20mt|MRP-L20|Mitochondrial large ribosomal subunit protein bL20m
- MRPL20|39S ribosomal protein L20|mitochondrial|L20mt|MRP-L20|Mitochondrial large ribosomal subunit protein bL20m
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYC9
- Q9BYC9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice