Anti-CHST9 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHST9
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-896
- 77829-895
- Antigen Symbol:
- CHST9
- CHST9
- Antigen Name:
- Carbohydrate sulfotransferase 9 protein (33-129AA)
- Carbohydrate sulfotransferase 9 protein (33-129AA)
- Antigen Synonyms:
- Carbohydrate sulfotransferase 9|Chst9|CHST9_HUMAN|GalNAc-4-O-sulfotransferase 2|GalNAc-4-ST2|GalNAc4ST-2|N-acetylgalactosamine-4-O-sulfotransferase 2|UNQ2549/PRO6175
- Carbohydrate sulfotransferase 9|Chst9|CHST9_HUMAN|GalNAc-4-O-sulfotransferase 2|GalNAc-4-ST2|GalNAc4ST-2|N-acetylgalactosamine-4-O-sulfotransferase 2|UNQ2549/PRO6175
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L1S5
- Q7L1S5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice