Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-AK3 Rabbit Recombinant Polyclonal Antibody
New Product
 
undefined
Anti-AK3 Rabbit Recombinant Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77841-561
  • 77841-562
  • Antigen Symbol:
  • AK3
  • AK3
  • Antigen Name:
  • GTP:AMP phosphotransferase AK3, mitochondrial protein (142-227AA)
  • GTP:AMP phosphotransferase AK3, mitochondrial protein (142-227AA)
  • Antigen Synonyms:
  • Adenylate kinase 3 alpha-like 1|Adenylate kinase 3|Adenylate kinase 3, formerly|adenylate kinase 6, adenylate kinase 3 like 1|AK 3|AK3, formerly|AK3L1|AK4|AK4, mouse, homolog of|AK6|AKL3L|AKL3L1|FIX|GTP:AMP phosphotransferase|GTP:AMP phosphotransferase, mitochondrial|KAD3_HUMAN|mitochondrial|OTTHUMP00000021015|OTTHUMP00000021016|RP11 6J24.4
  • Adenylate kinase 3 alpha-like 1|Adenylate kinase 3|Adenylate kinase 3, formerly|adenylate kinase 6, adenylate kinase 3 like 1|AK 3|AK3, formerly|AK3L1|AK4|AK4, mouse, homolog of|AK6|AKL3L|AKL3L1|FIX|GTP:AMP phosphotransferase|GTP:AMP phosphotransferase, mitochondrial|KAD3_HUMAN|mitochondrial|OTTHUMP00000021015|OTTHUMP00000021016|RP11 6J24.4
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UIJ7
  • Q9UIJ7
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice