Anti-Parvalbumin beta Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Parvalbumin beta
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gadus morhua subsp
Specifications
- Catalog No:
- 77817-836
- 77817-835
- Antigen Symbol:
- Parvalbumin beta
- Parvalbumin beta
- Antigen Name:
- Recombinant Gadus morhua subsp Parvalbumin beta protein (1-113AA)
- Recombinant Gadus morhua subsp Parvalbumin beta protein (1-113AA)
- Antigen Synonyms:
- Parvalbumin beta|Allergen Gad c I|Allergen M|allergen Gad c 1
- Parvalbumin beta|Allergen Gad c I|Allergen M|allergen Gad c 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Gadus morhua subsp
- Gadus morhua subsp
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02622
- P02622
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice