Anti-APOC4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: APOC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77814-245
- 77814-244
- Antigen Symbol:
- APOC4
- APOC4
- Antigen Name:
- Synthetic peptide of APOC4
- Synthetic peptide of APOC4
- Antigen Synonyms:
- APO C4|Apo CIV|Apo-CIV|APOC 4|ApoC IV|ApoC-IV|APOC4|APOC4_HUMAN|Apolipoprotein C IV|Apolipoprotein C-IV|Apolipoprotein C4
- APO C4|Apo CIV|Apo-CIV|APOC 4|ApoC IV|ApoC-IV|APOC4|APOC4_HUMAN|Apolipoprotein C IV|Apolipoprotein C-IV|Apolipoprotein C4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55056
- P55056
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice