Anti-ARHGAP32 Rabbit Recombinant Polyclonal Antibody
Catalog # 77818-194
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Rho GTPase-activating protein 32 protein (130-205AA)
- Antigen Symbol:ARHGAP32
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:A7KAX9
- Antigen Synonyms:ARHGAP32|GRIT|KIAA0712|RICS|Rho GTPase-activating protein 32|Brain-specific Rho GTPase-activating protein|GAB-associated Cdc42/Rac GTPase-activating protein|GC-GAP|GTPase regulator interacting with TrkA|Rho-type GTPase-activating protein 32|Rho/Cdc42/Rac GTPase-activating protein RICS|RhoGAP involved in the beta-catenin-N-cadherin and NMDA receptor signaling|p200RhoGAP|p250GAP
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:20 to 1:200
Type:
Antigen: ARHGAP32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human