Anti-ZNF408 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ZNF408
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-413
- 77805-412
- Antigen Symbol:
- ZNF408
- ZNF408
- Antigen Name:
- Zinc finger protein 408 protein (317-635AA)
- Zinc finger protein 408 protein (317-635AA)
- Antigen Synonyms:
- EVR6|PFM14|PR domain zinc finger protein 17|PRDM17|RP72|Zinc finger protein 408|ZN408_HUMAN|ZNF408
- EVR6|PFM14|PR domain zinc finger protein 17|PRDM17|RP72|Zinc finger protein 408|ZN408_HUMAN|ZNF408
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H9D4
- Q9H9D4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice