Anti-RAP2B Rabbit Polyclonal Antibody
77796-657
New Product
- Antigen Name:Small GTP binding protein
- Antigen Symbol:RAP2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P61225
- Antigen Synonyms:MGC20484|RAP 2B|RAP2A|Rap2b|RAP2B member of RAS oncogene family|RAP2B_HUMAN|Ras family small GTP binding protein RAP2B|Ras related protein RAP 2B|Ras related protein RAP2B|Ras-related protein Rap-2b|Small GTP binding protein
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: RAP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat