Anti-ALG13 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ALG13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-266
- 77842-267
- Antigen Symbol:
- ALG13
- ALG13
- Antigen Name:
- Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13 protein (112-285AA)
- Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13 protein (112-285AA)
- Antigen Synonyms:
- ALG13|ALG13, S. cerevisiae, homolog of|ALG13, UDP-N-acetylglucosaminyltransferase subunit|ALG13_HUMAN|Asparagine linked glycosylation 13 homolog (S. cerevisiae)|Asparagine-linked glycosylation 13 homolog|Asparagine-linked glycosylation 13, S. cerevisiae, homolog of|CDG1S|CXorf45|FLJ23018|FLJ31785|GLT28D1|Glycosyltransferase 28 domain containing protein 1|Glycosyltransferase 28 domain-containing 1|Glycosyltransferase 28 domain-containing protein 1|Hematopoietic stem/progenitor cells protein MDS031|MDS031|MGC12423|N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase|OTTHUMP00000062812|OTTHUMP00000176862|TDRD13|tudor domain containing 13|UDP N acetylglucosamine transferase subunit ALG13 homolog|UDP-N-acetylglucosamine transferase subunit ALG13 homolog|YGL047W
- ALG13|ALG13, S. cerevisiae, homolog of|ALG13, UDP-N-acetylglucosaminyltransferase subunit|ALG13_HUMAN|Asparagine linked glycosylation 13 homolog (S. cerevisiae)|Asparagine-linked glycosylation 13 homolog|Asparagine-linked glycosylation 13, S. cerevisiae, homolog of|CDG1S|CXorf45|FLJ23018|FLJ31785|GLT28D1|Glycosyltransferase 28 domain containing protein 1|Glycosyltransferase 28 domain-containing 1|Glycosyltransferase 28 domain-containing protein 1|Hematopoietic stem/progenitor cells protein MDS031|MDS031|MGC12423|N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase|OTTHUMP00000062812|OTTHUMP00000176862|TDRD13|tudor domain containing 13|UDP N acetylglucosamine transferase subunit ALG13 homolog|UDP-N-acetylglucosamine transferase subunit ALG13 homolog|YGL047W
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NP73
- Q9NP73
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice